Anti-PDPK1 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

600-401-411
ROCK600-401-411EA 6120 DKK
ROCK600-401-411
Anti-PDPK1 Rabbit Polyclonal Antibody
Antistoffer
This affinity purified antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band ~60 kDa in size corresponding to human PDK-1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR