Anti-NFYA Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

200-401-100
ROCK200-401-100EA 6120 DKK
ROCK200-401-100
Anti-NFYA Rabbit Polyclonal Antibody
Antistoffer
Anti-NF-Y antibody is suitable for western blotting and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000;

Type: Primary
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR