Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 18F3.H11]

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

200-341-269
ROCK200-341-269EA 6030 DKK
ROCK200-341-269
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 18F3.H11]
Antistoffer
This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:4000-1:20000; FLISA: >1:20000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 18F3.H11
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey
Bestil nu

Specifikationer

Antigen symbol AKT1
Environmentally Preferable
Antigen name v-akt murine thymoma viral oncogene homolog 1
Antigen synonymer CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
Antistof type Primary
Clonality Monoclonal
Konjugation DyLight® 488
Clone 18F3.H11
Reactivity Monkey,Human,Rat,Mouse
Vært Mouse
Gen ID 207
Isotype IgG1 kappa
Western blot Yes
ELISA Yes
ImmunFluorescens Yes
Flowcytometri Yes
Modification phospho-Thr308
Immunogen This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding T308 of human AKT1 Protein.
Udformning Antibody fraction
Form Lyophilized
Purification Protein A purified
Krydsadsorption No
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Opbevaringstemperatur –20 °C
Shipping temperature Ambient
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR