Anti-HRP Rat Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100-5138
ROCK100-5138EA 5090 DKK
ROCK100-5138
Anti-HRP Rat Polyclonal Antibody
Antistoffer
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:2000 - 1:6000; Western Blot: 1:200 - 1:1000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: pan
Bestil nu

Specifikationer

Environmentally Preferable
Antigen name Horseradish Peroxidase
Antistof type Tag
Clonality Polyclonal
Konjugation Unconjugated
Reactivity Multiple
Vært Rat
Isotype
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Immunogen Peroxidase (Horseradish)
Udformning Antiserum
Form Lyophilized
Purification delipidation and defibrination
Koncentration 85 mg/ml by Refractometry
Krydsadsorption none
Storage buffer 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2,
Opbevaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR