Anti-FBXO4 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100-401-963
ROCK100-401-963EA 6120 DKK
ROCK100-401-963
Anti-FBXO4 Rabbit Polyclonal Antibody
Antistoffer
This antiserum has been tested for use in western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 – 1:3000;

Type: Primary
Antigen: FBXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestil nu

Specifikationer

Antigen symbol FBXO4
Environmentally Preferable
Antigen name F-box protein 4
Antigen synonymer AW494535|AI851261|1700096C12Rik|Fbx4
Antistof type Primary
Clonality Polyclonal
Konjugation Unconjugated
Reactivity Human,Mouse
Vært Rabbit
Gen ID 106052
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitop N-terminal
Immunogen This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide derived from sequences unique to the N-terminus of FBX4 and conserved between the human and murine Proteins (see link below for the full-length sequence of the mouse g
Udformning Antiserum
Form Liquid (sterile filtered)
Krydsadsorption No
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
Opbevaringstemperatur –20 °C
Shipping temperature Dry Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR