Bottles, two necks, rectangular, HDPE
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3-5100-1000-28
RIXI3-5100-1000-28EA 2460 DKK
RIXI3-5100-1000-28
Bottles, two necks, rectangular, HDPE
Flasker Plastflasker
Two-neck bottle made up of HDPE, natural.
  • Rigid and shatter-proof
  • Resistant to water, aqueous solutions and most chemicals
  • Excellent water vapour resistance
Bestillingsoplysninger: Screw caps must be ordered seperately.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904