Universal General Purpose Ultra-Low Freezers, TSX
Leverandør: REVCO TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TSX40086FA TSX50086FA TSX60086FA TSX70086FA
REVCTSX40086FAEA 0 DKK
REVCTSX40086FA REVCTSX50086FA REVCTSX60086FA REVCTSX70086FA
Universal General Purpose Ultra-Low Freezers, TSX
Frysere Ultra lav temperatur frysere
The TSX Universal heralds a new era in performance with Universal V-Drive technology, delivering tighter control and faster recovery times. Coupled with an expanded setpoint range, universal voltage, whisper-quiet operation, simplified software updates and much more, TSX Universal seamlessly adapts to your workflow.
  • Ensure reliability and security for your valuable samples
  • Designed to meet diverse research demands
  • TSX universal ULT redefines standards with its next-gen features
Redefining sustainability, it is ENERGY STAR certified with a 32%More Product Information