PBDE mix (5 components)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
33098
REST33098EA 8060 DKK
REST33098
PBDE mix (5 components)
Standarder Økologiske standarder
The PBDE mix consists of five components, they are 2,2',4,4',5,5'-hexabromobiphenyl, 2,2',4,4',5,5'-Hexabromodiphenyl ether (BDE-153), 2,2',4,4',5-Pentabromodiphenyl ether (BDE-99) , 2,2',4,4',6-Pentabromodiphenyl ether (BDE-100) and 2,2',4,4'-Tetrabromodiphenyl ether (BDE-47).