CLP 04.1 B/N MegaMix standard, revision 2 (47 components)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31903
REST31903EA 1330 DKK
REST31903
CLP 04.1 B/N MegaMix standard, revision 2 (47 components)
Standarder Økologiske standarder
Consists of 47 components, those are acenaphthene, acenaphthylene, acetophenone, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, benzyl butyl phthalate, biphenyl, bismethane, bisether, bisphthalate, 4-bromophenyl phenyl ether, carbazole, 4-chloroaniline, 2-chloronaphthalene, 4-chlorophenyl phenyl ether, chrysene, dibenzanthracene, dibenzofuran, diethylphthalate, dimethylphthalate, di-n-butyl phthalate, 2,4-dinitrotoluene, 2,6-dinitrotoluene, di-n-octyl phthalate, diphenylamine *,fluoranthene, fluorene, hexachlorobenzene, hexachloro-1,3-butadiene , hexachlorocyclopentadiene, hexachloroethane, indenopyrene, isophorone, 2-methylnaphthalene, naphthalene, 2-nitroaniline, 3-nitroaniline, 4-nitroaniline, nitrobenzene, n-nitroso-di-n-propylamine, 2,2'-oxybis, phenanthrene and pyrene.
Certificeringer: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Bestillingsoplysninger: *N-nitrosodiphenylamine (CLP-listed analyte) decomposes to diphenylamine (mix component) in the injector.