MA fractionation surrogate spike mix (2 components)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31480
REST31480EA 366 DKK
REST31480
MA fractionation surrogate spike mix (2 components)
Standarder Miljø analyse standarder
This MA fractionation surrogate spike mix contains two components: 2-Bromonaphthalene and 2-Fluorobiphenyl.
Shelf life 6 months.
Certificeringer: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.