Flow controllers, Veriflo
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24218 24219 24229 24232 24233 24234 24235 24236 24237 24238 24239 24245 24246 24247 24248 24249 24255 24256 24257 24258 24259 24260 24261 24262 24263 24264
REST24218EA 1450 DKK
REST24218 REST24219 REST24229 REST24232 REST24233 REST24234 REST24235 REST24236 REST24237 REST24238 REST24239 REST24245 REST24246 REST24247 REST24248 REST24249 REST24255 REST24256 REST24257 REST24258 REST24259 REST24260 REST24261 REST24262 REST24263 REST24264
Flow controllers, Veriflo
Flow switches
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.