Valves and loops, Sulfinert® treated
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20584 20585 20586
REST20584EA 21480 DKK
REST20584 REST20585 REST20586
Valves and loops, Sulfinert® treated
Ventiler Injektionsventiler
Sulfinert® treated gas sample loops are ideal for samples containing low concentrations of sulphur and other active compounds.