Calciumchlorid dihydrat

Leverandør: G Biosciences
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Calcium Chloride Dihydrate, Calcium dichloride

RC-030
RC-030EA 973 DKK
RC-030
Calciumchlorid dihydrat
Calciumchlorid dihydrat
Formel: CaCl₂·2H₂O
Molekylvægt: 147.01 g/mol
Kogepunkt: 1600
Smeltepunkt: 176
Massefylde: 1.85 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149613
CAS-nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR