Pipette tips, ratiolab®
Leverandør: RATIOLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2500190 2500170 2500160 2500110 2400630 2400620 2400600 2300650 2400610 2400640 2100610 2100600 2400650 2500130 2513163 2410650 2113602 2530162 2410630 2130602 2530172 2400690 2153602 2500163 2513172 2530173 2100500 2113612 2530192 2513502 2553502 2500180 2410620 2413602 2453602 2153612 2413612 2400622 2400624 2430602 2513162 2553162 2453612 2430612 2400623
612-1476EA 230 DKK
612-1476 612-1477 612-1478 612-1480 612-1483 612-1484 612-1487 612-1488 612-1489 612-1490 612-2501 612-2502 RATI2400650 RATI2500130 RATI2513163 RATI2410650 RATI2113602 RATI2530162 RATI2410630 RATI2130602 RATI2530172 RATI2400690 RATI2153602 RATI2500163 RATI2513172 RATI2530173 RATI2100500 RATI2113612 RATI2530192 RATI2513502 RATI2553502 RATI2500180 RATI2410620 RATI2413602 RATI2453602 RATI2153612 RATI2413612 RATI2400622 RATI2400624 RATI2430602 RATI2513162 RATI2553162 RATI2453612 RATI2430612 RATI2400623
Pipette tips, ratiolab®
Pipettespidser
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
  • Made of high quality PP
  • Tight fit due to the precisely fitting cone
  • Guaranteed dust-free due to special ionization treatment
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.