Kaliumhydroxid 2 mol/l (2 N) aqueous solution

Leverandør: G Biosciences
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potash caustic, Caustic potash

R008
R008EA 594 DKK
R008
Kaliumhydroxid 2 mol/l (2 N) aqueous solution
Potassium hydroxide
Formel: KOH
Molekylvægt: 56.11 g/mol
Kogepunkt: 1320
Smeltepunkt: 360
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
UN nummer: 1814
ADR: 8 III
Merck Index: 12,07806

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR