Anti-Quinidine Mouse Monoclonal Antibody

Leverandør: QED Bioscience
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

16502
QEDB16502EA 3030 DKK
QEDB16502
Anti-Quinidine Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Quinidine
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR