Disposable particle filtering respirators, FFP2, P251
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P251WHR
PRTWP251WHREA 206 DKK
PRTWP251WHR
Disposable particle filtering respirators, FFP2, P251
Åndedrætsværn Engangsmasker Partikelmasker
Vertical flat folded mask with exhalation valve for lighter breathing resistance. Ideal for all day use.
  • Adjustable nose clip for optimised fit
  • Ultrasound welded elastic bands
  • Flexible shape allows the mask to be folded flat
  • Individually packed for vending machines
Certificeringer: EN 149:2001 + A1:2009.
Emballering: Individually wrapped inside hygienic polybag.