Waist wader, Steelite™ Safety, FW71
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FW71BKR37 FW71BKR38 FW71BKR39 FW71BKR40 FW71BKR41 FW71BKR42 FW71BKR43 FW71BKR44 FW71BKR45 FW71BKR46 FW71BKR47
PRTWFW71BKR37EA 422 DKK
PRTWFW71BKR37 PRTWFW71BKR38 PRTWFW71BKR39 PRTWFW71BKR40 PRTWFW71BKR41 PRTWFW71BKR42 PRTWFW71BKR43 PRTWFW71BKR44 PRTWFW71BKR45 PRTWFW71BKR46 PRTWFW71BKR47
Waist wader, Steelite™ Safety, FW71
Arbejdstøj
Heavy-duty PVC/nitrile safety waist wader with adjustable thigh straps and integrated S5 safety boots.
  • Colour-coded red outsole for easy identification of safety features
  • Slip-and oil resistant, anti-static outsole
  • Energy absorber heel
  • Toe protection cap made from steel (200 J)
  • Pierce resistant steel midsole
Colour: Black
EN ISO 20345:2011, S5
Certificeringer: EN ISO 20345:2011, S5