Safety boots, pull-on, Steelite™ WORK Rigger FW12
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FW12TAR44 FW12TAR47 FW12TAR46 FW12TAR43 FW12TAR45 FW12BKR38 FW12TAR42 FW12TAR41 FW12TAR40 FW12TAR39 FW12TAR38 FW12BKR47 FW12BKR46 FW12BKR45 FW12BKR44 FW12BKR43 FW12BKR42 FW12BKR41 FW12BKR39 FW12BKR40
PRTWFW12TAR44EA 512 DKK
PRTWFW12TAR44 PRTWFW12TAR47 PRTWFW12TAR46 PRTWFW12TAR43 PRTWFW12TAR45 PRTWFW12BKR38 PRTWFW12TAR42 PRTWFW12TAR41 PRTWFW12TAR40 PRTWFW12TAR39 PRTWFW12TAR38 PRTWFW12BKR47 PRTWFW12BKR46 PRTWFW12BKR45 PRTWFW12BKR44 PRTWFW12BKR43 PRTWFW12BKR42 PRTWFW12BKR41 PRTWFW12BKR39 PRTWFW12BKR40
Safety boots, pull-on, Steelite™ WORK Rigger FW12
Sko og støvler
Anti-static pull-on boots made from split buffalo leather plus a dual-density, PU/rubber sole resistant to oil and heat (300 °C).
  • Fur lining
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
  • Side tabs for easy donning
Colour: Black/brown
Certificeringer: EN ISO 20345:2011; S1P, CI, HRO, SRC.