Tin standard solution, for ICP, in HCl, CRM, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHG-PSNH-100 VHG-LSNH-100 VHG-PSNH-50 VHG-PSNH-500 VHG-TSNH-100 VHG-TSNH-500
PROOVHG-PSNH-100EA 632 DKK
PROOVHG-PSNH-100 PROOVHG-LSNH-100 PROOVHG-PSNH-50 PROOVHG-PSNH-500 PROOVHG-TSNH-100 PROOVHG-TSNH-500
Tin standard solution, for ICP, in HCl, CRM, VHG
Standarder Kationiske standarder Tin Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificeringer: Traceable to NIST, SRM.