Potassium standard solution, for ICP, in 5% HNO₃, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHG-PKN-100 VHG-PKN-500 VHG-TKN-100 VHG-TKN-500 VHG-PKN-250
PROOVHG-PKN-100EA 620 DKK
PROOVHG-PKN-100 PROOVHG-PKN-500 PROOVHG-TKN-100 PROOVHG-TKN-500 PROOVHG-PKN-250
Potassium standard solution, for ICP, in 5% HNO₃, VHG
Standarder Kationiske standarder Potassium Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificeringer: Traceable to NIST, SRM.