Tungsten standard solution, for ICP, in 5% HNO₃, tr. HF, CRM, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHG-PWNF-100 VHG-PWNF-500 VHG-TWNF-100 VHG-TWNF-500
PROOVHG-PWNF-100EA 632 DKK
PROOVHG-PWNF-100 PROOVHG-PWNF-500 PROOVHG-TWNF-100 PROOVHG-TWNF-500
Tungsten standard solution, for ICP, in 5% HNO₃, tr. HF, CRM, VHG
Standarder Kationiske standarder Tungsten Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificeringer: Traceable to NIST, SRM.