Methyl-4-aminosalicylate

Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 4-amino-2-hydroxybenzoate

MM0073.09
PROOMM0073.09EA 0 DKK
PROOMM0073.09
Methyl-4-aminosalicylate
Methyl-4-aminosalicylate
Formel: C₈H₉NO₃
MDL Number: MFCD00088091
CAS-nummer: 4136-97-4
EINECS: 223-958-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR