Imject™ Bovine serum albumin (BSA)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
77110
PIER77110EA 2210 DKK
PIER77110
Imject™ Bovine serum albumin (BSA)
Proteiner
Imject™ BSA in phosphate buffered saline (PBS) is purified bovine serum albumin carrier protein that enables simple preparation of effective immunogens with amine- or carboxyl-peptide antigens.
  • 5×20 mg