Protein L Plus agarose, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20520
PIER20520EA 4760 DKK
PIER20520
Protein L Plus agarose, Pierce™
Protein oprensnings Protein Purification Resins
Pierce™ Protein L Plus agarose is a high-capacity Protein L beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilised Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein L is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • High capacity beads with a dense load of immobilised Protein L
Binding capacity: 10 to 20 mg human IgG/ml resin