Immunoassay microplates, biotin coated, 96-well strip plates, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
15151
PIER15151EA 2000 DKK
PIER15151
Immunoassay microplates, biotin coated, 96-well strip plates, Pierce™
Mikroplader Immunologi mikroplader
Biotin coated plates can be used as the basis for a variety of biomolecular assay techniques requiring the immobilisation of probes that are conjugated with streptavidin, avidin, or other biotin-binding proteins.
  • Biotin group accessible for binding avidin or streptavidin
  • Pre-blocked to reduce non specific binding
  • Strip well plate format for the ultimate in convenience
  • Coat volume: 200 µl
  • Blocking volume: 300 µl