Foaming Solutions for Hygienic Hand Washing

Leverandør: PHAGOGENE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

65123 SA0141050
PHAG65123EA 523 DKK
PHAG65123 PHAGSA0141050
Foaming Solutions for Hygienic Hand Washing
Hudpleje Sæber og huden rensemidler
Liquid washing and disinfecting soap specially formulated for the hygienic hands washing.

  • Skin-friendly formula even in case of frequent washing
  • Leave your hands soft
  • Enriched with glycerine
  • Without perfume

Used before any aseptic gesture (catheters and more) and systematically before and after any contact with a patient or contaminating material. Specially formulated for the medical, food and institutional sectors.

Appearance: Gelled liquid, opalescent
Colour: Colourless
Odor: Alcohol
pH at 20 °C (pure product): 7,0 ±0,5
Density at 20 °C (g/ml): 0,905More Product Information
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR