Capillary columns, Elite-Alumina for autosystem
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9316544 N9316304 N9316305
PERCN9316544EA 41550 DKK
PERCN9316544 PERCN9316304 PERCN9316305
Capillary columns, Elite-Alumina for autosystem
Kromatografikolonner
Elite-Alumina PLOT phase capillary columns are ideal for the analysis of light hydrocarbons.