Capillary columns, Elite-225 for autosystem
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9316175 N9316177 N9316181
PERCN9316175EA 7690 DKK
PERCN9316175 PERCN9316177 PERCN9316181
Capillary columns, Elite-225 for autosystem
Kromatografikolonner
Elite-225 columns are fused silica columns coated with phenylmethylpolysiloxane (50% cyanopropylphenyl).