Gas purification systems
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9303963 N9303964 N9306139
PERCN9303963EA 24160 DKK
PERCN9303963 PERCN9303964 PERCN9306139
Gas purification systems
Filtre Gasfiltre
Moisture, oxygen, and other trace contaminants found in carrier or make-up gases can greatly affect your analysis and instrument performance. They can cause ghost peaks and degrade your column. Gas filters and traps can remove up to 99,9999% of these unwanted contaminants, increasing the life of your column life and decreasing stationary phase bleed.
  • Easy cartridge replacement with on/off knob
  • Double seal construction for safety
  • Two indicators for both oxygen and moisture
  • High capacity and efficiency levels for oxygen, water and hydrocarbons