Pyridin ≥99%, Reagent Grade

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Azabenzene, Azine

P57506-1L P57506-2.5L P57506-250ML P57506-500ML
P57506-1LEA 1220 DKK
P57506-1L P57506-2.5L P57506-250ML P57506-500ML
Pyridin ≥99%, Reagent Grade
Pyridine

  • ≥95% for general laboratory use.

Formel: C₅H₅N
Molekylvægt: 79.1 g/mol
Kogepunkt: 12.4 %(V)
Smeltepunkt: –42 °C
Massefylde: 0.98 g/cm³ (20 °C)
Flammepunkt: 20 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011732
CAS-nummer: 110-86-1
EINECS: 203-809-9
UN nummer: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Autoignition Temperature 899 °F
IR spectrum complying
Assay (GC) Min. 99.0 %
Assay (HClO₄-titr.) 99.0 - 101.0 %
Water (Karl Fischer) Max. 0.1 %
APHA Max. 50
Vapor Pressure 10 mmHg ( 13.2 °C)
Refractive Index n20/D 1.509 (lit.)
Explosion Limit 12.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR