Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P0425-72EA
P0425-72EAEA 896 DKK
P0425-72EA
Objektglas Adhæsionsobjektglas
Poly-prep slides are made of glass and designed for cell adhesion. The slides are coated with Poly-L-Lysine, a non-specific attachment factor for cells useful in promoting cell adhesion on solid substrates. Poly-L-lysine enhances binding between the cell membrane and the cultured surface.
  • Poly-Prep Slides are used:
  • To mount liver and lung cell sections and mounted on poly-prep slides
  • To prove better cell adhesion in clinical bronchoalveolar lavage fluid samples
  • To determine pAkt (alpha serine/threonine-protein kinase) expression by immunohistochemistry
  • In