DNA transfection reagent, PolyMag™
Leverandør: OZ BIOSCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PN30200 PN31000 PN30100
OZBIPN30200EA 1810 DKK
OZBIPN30200 OZBIPN31000 OZBIPN30100
DNA transfection reagent, PolyMag™
Transfektionsreagenser
PolyMag™ transfection reagent, based on Magnetofection™ transfection technology, has been optimised for use with all types of nucleic acids. The reagent is composed of magnetic nanoparticles coated with cationic molecules. It has been successfully tested on a large range of primary cells, hard to transfect cells and cell lines and its efficiency has been reported in many publications.
  • Highly efficient for primary cells
  • Powerful for hard to transfect cells
  • Non toxic
  • Serum-compatible
  • Simple, rapid and easy to use
  • For all types of nucleic acids
Store at +4°C.
Information om tilbehør: This reagent needs to be used with a magnetic plate.
Leveringsinformation: Shipped at room temperature.