Wafer tweezers
Leverandør: OUTILS RUBIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37S-2 37S-1
OUTI37S-2EA 389 DKK
OUTI37S-2 OUTI37S-1
Wafer tweezers
Pincet
Stainless steel. These narrow tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
  • Stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping
  • Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
  • Satin finish on the outside of the paddel for glare prevention
  • Each Wafer size has its corresponding tweezers size (e.g. for an 8" Wafer take tweezers with a No. 8)