Ion selective electrode, half-cell, fluoroborate, Orion™
Leverandør: Thermo Orion
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9305BN
ORIO9305BNEA 24040 DKK
ORIO9305BN
Ion selective electrode, half-cell, fluoroborate, Orion™
Elektroder ISE elektroder
This fluoroborate half-cell can be easily and economically rejuvenated.
Plastic membrane half-cell requires reference electrode.