Multifunctional standard bench scales, Defender 5000
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30461581 30461582 30461583 30461584 30461585 30461586 30461587 30461588 30461589 30461590 30461591 30461551 30461552 30461553 30461554 30461555 30424312 30424313 30424314 30424315 30424316 30424317 30424318 30424319 30424320 30424321 30424322 30424323 30424324 30461545 30461546 30461547 30461548 30461549 30461550 30461556 30461557 30444804 30444805 30444806 30444807 30444808 30444809 30444810 30444811 30444812 30444813 30444814 30444815 30444816
OHAU30461581EA 11790 DKK
OHAU30461581 OHAU30461582 OHAU30461583 OHAU30461584 OHAU30461585 OHAU30461586 OHAU30461587 OHAU30461588 OHAU30461589 OHAU30461590 OHAU30461591 OHAU30461551 OHAU30461552 OHAU30461553 OHAU30461554 OHAU30461555 611-4921 611-4922 611-4923 611-4924 611-4925 611-4926 611-4927 611-4928 611-4929 611-4930 611-4931 611-4932 611-4933 OHAU30461545 OHAU30461546 OHAU30461547 OHAU30461548 OHAU30461549 OHAU30461550 OHAU30461556 OHAU30461557 OHAU30444804 OHAU30444805 OHAU30444806 OHAU30444807 OHAU30444808 OHAU30444809 OHAU30444810 OHAU30444811 OHAU30444812 OHAU30444813 OHAU30444814 OHAU30444815 OHAU30444816
Multifunctional standard bench scales, Defender 5000
Vægte
Defender 5000 series multifunctional bench scales are ideal for numerous applications including production, packaging, inventory and shipping. Durably constructed to withstand harsh environments and equipped with a stainless steel platform, powder coated steel frame and an aluminium load cell. Choice of indicator: D52P unit is ABS plastic and the D52XW indicator is 304 stainless steel with sand blasted surface treatment, which offers protection to IP 68.
  • Large backlit display and alphanumeric keypad with function keys
  • Indicator guides user with on-screen