Multifunctional standard front mounted bench scales Defender 5000
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30461610 30461611 30461612 30461613 30461614 30461615 30461616 30424325 30424326 30424327 30444761 30444762 30444763 30444764 30461558 30461559 30461560 30461561 30461562 30461563 30461564 30498216 30498217 30498498 30498499 30498500 30498501 30498502
OHAU30461610EA 11740 DKK
OHAU30461610 OHAU30461611 OHAU30461612 OHAU30461613 OHAU30461614 OHAU30461615 OHAU30461616 OHAU30424325 OHAU30424326 OHAU30424327 OHAU30444761 OHAU30444762 OHAU30444763 OHAU30444764 OHAU30461558 OHAU30461559 OHAU30461560 OHAU30461561 OHAU30461562 OHAU30461563 OHAU30461564 OHAU30498216 OHAU30498217 OHAU30498498 OHAU30498499 OHAU30498500 OHAU30498501 OHAU30498502
Multifunctional standard front mounted bench scales Defender 5000
Vægte
The Defender 5000 Series multifunctional bench scales are ideal for numerous applications including production, packaging, inventory and shipping. Durably constructed to withstand harsh environments and equipped with a stainless steel platform, powder-coated steel frame and an aluminium load cell. Choice of indicator: D52P unit is ABS plastic and the D52XW indicator is 304 stainless steel with sand blasted surface treatment, which offers protection to IP 68.
  • Large backlit display and alpha-numeric keypad with function keys
  • Indicator guides user with on-