Anti-SARS-CoV-2 nsp12 Rabbit Monoclonal Antibody [clone: HL1444]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP3-25653
NOVUNBP3-25653EA 5390 DKK
NOVUNBP3-25653
Anti-SARS-CoV-2 nsp12 Rabbit Monoclonal Antibody [clone: HL1444]
Antistoffer
  • Immunocytochemistry/Immunofluorescence: Assay dependent
  • Immunohistochemistry: Assay dependent
  • Immunohistochemistry-Paraffin: Assay dependent
  • Western blot: 1:1000 - 1:10000

Advarsel: Research use only.

Type: Primary
Antigen: SARS-CoV-2 nsp12
Clonality: Monoclonal
Clone: HL1444
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR