Anti-CCL2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NBP1-07035F
NOVUNBP1-07035FEA 5390 DKK
NOVUNBP1-07035F
Anti-CCL2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR