Rabbit Polyclonal anti-160kDa Neurofilament Medium antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NB300-133
NOVUNB300-133EA 4470 DKK
NOVUNB300-133
Rabbit Polyclonal anti-160kDa Neurofilament Medium antibody
Antistoffer
The NF-M Antibody from Novus Biologicals is a rabbit polyclonal antibody to NF-M. This antibody reacts with human, mouse, rat, porcine, avian, bovine, feline. The NF-M Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000

Type: Primary
Antigen: NF-M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Avian, Bovine, Feline
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR