Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 405)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NB110-40765V
NOVUNB110-40765VEA 4990 DKK
NOVUNB110-40765V
Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 405)
Antistoffer
Type: Primary
Antigen: PLIN3 (perilipin 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR