Anti-5-HT3A Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NB100-56351
NOVUNB100-56351EA 4500 DKK
NOVUNB100-56351
Anti-5-HT3A Rabbit Polyclonal Antibody
Antistoffer
The 5-HT3A Antibody from Novus Biologicals is a rabbit polyclonal antibody to 5-HT3A. This antibody reacts with human, mouse, rat. The 5-HT3A Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Simple Western,

Type: Primary
Antigen: 5-HT3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Specifikationer

Antigen symbol 5HT3
Environmentally Preferable
Antigen name 5-hydroxytryptamine receptor 3a
Antigen synonymer 5-HT3A|5HT3 receptor|5-HT3|5 HT3|5HT3|79246|Htr3|5 HT3A|Htr3a|5HT3A|5 HT3 receptor|5HT
Antistof type Primary
Clonality Polyclonal
Konjugation Unconjugated
Reactivity Human
Vært Rabbit
Gen ID 3359
Isotype
Western blot Yes
Storage buffer PBS
Opbevaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR