Cleanroom face masks, BioClean™ MHA
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MHA210-0
NITRMHA210-0EA 0 DKK
NITRMHA210-0
Cleanroom face masks, BioClean™ MHA
Masker Renrumsmasker
These BioClean MHA™ cleanroom headloop facemasks are designed to meet the demands of the most exacting operating environments.
  • Width: 210 mm to provide superior protection
  • Clearly marked to differentiate inner and outer surface
  • Extra length conformable noseband ensures wearer comfort
  • Latex-free
Non sterile layers: Inner layer non-woven PP/PE (hygroscopic), filter layer meltblown PP, outer layer non-woven PP/PE (hydrophobic), headloops PU.
Sterile layers: Inner layer non-woven PP/PE (hydrophilic), filter layer meltblown PP, outer layer non-woven PP/PE (More Product Information