Sludge Judge® sampling tools
Leverandør: NASCO FORT ATKINSON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C09247WA B01355WA
NASCC09247WAEA 2360 DKK
NASCC09247WA NASCB01355WA
Sludge Judge® sampling tools
Prøvetagere Væskeprøvetagere
The Sludge Judge line of collection tools are dependable and long-lasting.
  • Designed to take accurate readings of settleable solids
  • Ideal for anywhere that accurate sample levels of settleable solids in noncaustic materials are needed
  • Maximum strength and rigidity
  • Treated to reduce deterioration from sun's harmful rays
  • Durable in cold temperature
  • Float valve for easy sample collection and release
Do NOT use for sampling ethanol or ethanol blended fuel. One-year warranty.
To use:
1. Slowly lower the Sludge Judge® to the bottom of the tank. The floatMore Product Information