Bottle carriers, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6565-0001
NALG6565-0001EA 2590 DKK
NALG6565-0001
Bottle carriers, Nalgene®
Bærere Flaskebærere
These bottle carriers are versatile, durable and lightweight with partitions and assembly instruction printed on each tote.
HDPE, blue.
  • Holds upto 12,7 kg
  • Stores flat and easy to assemble
  • Carrier is not autoclavable