Nalgene® Media Bottles, Square, Amber, with Screw Cap
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
322021-0125 322021-0030 322021-0060
NALG322021-0125EA 1970 DKK
NALG322021-0125 NALG322021-0030 NALG322021-0060
Nalgene® Media Bottles, Square, Amber, with Screw Cap
Flasker Medieflasker
PETG, amber, translucent, with HDPE screw cap.
  • Non-sterile
  • Non-pyrogenic
  • Non-cytotoxic
Ideal for storage and shipping of light sensitive liquid media, buffers, and sera. Heavy-walled bottle provides an excellent gas barrier; features molded-in graduations, with a drip-proof neck finish and molded-in neck ring.
Heavy-walled, shatter resistant and durable, space-saving square design, molded in graduations, nonsterile, non-pyrogenic, non-cytotoxic, leakproof, with drip-proof neck finish and molded-in neck ring, PETG provides excellent gas barrier,More Product Information