Natriumchlorit, high purity

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

N819-100GEA 1040 DKK
N819-100G
Natriumchlorit, high purity
Sodium chlorite
Formel: ClNaO₂
Molekylvægt: 90.44 g/mol
Kogepunkt: 112
Smeltepunkt: 250…257 °C
Massefylde: 1.28 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003478
CAS-nummer: 7758-19-2
EINECS: 231-836-6
UN nummer: 1908
ADR: 8 II
Merck Index: 13,08672

Bestil nu

Specifikationer Testresultater

Arsenic <3 ppm
Heavy Metals (as Pb) <20 ppm
Hydrogen Peroxide 0.01 %
Lead <10 ppm
Mercury <0.1 ppm
Sodium Carbonate <2 %
Sodium Chlorate <4%
Sodium Chloride 11 – 19 %
Sodium Chlorite 77.5 – 82.5 %
Sodium Hydroxide <3%
Sodium Sulphate <3%
Water (by difference) <6%

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR