Oil sorbents socks
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9930 9931
MSGA9930EA 684 DKK
MSGA9930 MSGA9931
Oil sorbents socks
Spild materialer
Water repellent absorbent socks for safe absorption of oil and petrol.
  • Creates an absorption barrier to enclose larger leaks
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance