Module for Remote Signalling
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
38765 39221
MSGA38765EA 2920 DKK
MSGA38765 MSGA39221
Module for Remote Signalling
Møbler Opbevaringskabinetter
The module offers an alarm forwarding via the mobile phone network (SMS/call) and is therefore especially suitable for companies and facilities without a central building management system. In an emergency, the immediate alerting of one or several defined persons is triggered.
  • The module is easily installable on-site via plug
  • Five programmable telephone numbers
  • Configurable message texts
  • Collective alarm in case of an emergency or power failure
A SIM card is required for the initial operation of the module, which must be provided by the customer.More Product Information