Hygiene monitoring and management system, Clean-Trace™
Leverandør: 3M Food Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ATP10
MMMPATP10EA 711 DKK
MMMPATP10
Hygiene monitoring and management system, Clean-Trace™
Overvågningssystem Hygiejne Monitoreringssystemer
The Clean-Trace™ luminometer is a simple, quick and reliable handheld diagnostic tool to verify cleaning effectiveness and capture, store and manage data for later analysis. This Hygiene Monitoring and Management System offers food safety professionals a complete system of products consisting of a luminometer handheld device, compatible tests and a powerful new software platform, to verify that surfaces have been effectively cleaned in seconds, for better decision making and audit preparedness.
  • Improved precision and accuracy
  • Robust for the manufacturing environment
  • Ergonomic design